missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-(4-Chlorophenyl)ethylamine, ChiPros™ 97%, ee 98%

Catalog No. AAL1911914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1911914 25 g
AAL1911903 1 g
AAL1911906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1911914 Supplier Thermo Scientific Chemicals Supplier No. L1911914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 27298-99-3 | C8H10ClN | 155.625 g/mol

(R)-(+)-1-(4-Chlorophenyl)ethylamine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as a chiral reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-1-(4-Chlorophenyl)ethylamine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as a chiral reagent.

Solubility
Soluble in DMSO, insoluble in water.

Notes
Air Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 27298-99-3
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD00671639
InChI Key PINPOEWMCLFRRB-ZCFIWIBFSA-N
Synonym r-1-4-chlorophenyl ethylamine, r-1-4-chlorophenyl ethanamine, 1r-1-4-chlorophenyl ethanamine, r-+-1-4-chlorophenyl ethylamine, 1r-1-4-chlorophenyl ethan-1-amine, benzenemethanamine, 4-chloro-alpha-methyl-, alphar, pubchem15222, r-4-chloro-, a-methylbenzylamine, 1r-1-4-chlorophenyl ethylamine, r-1-4-chloro-phenyl ethylamine
PubChem CID 1715226
IUPAC Name (1R)-1-(4-chlorophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)Cl)N

Specifications

Density 1.11
Quantity 25 g
Refractive Index 1.5418
UN Number UN2922
Beilstein 2412317
Boiling Point 232°C
Solubility Information Soluble in DMSO,insoluble in water.
Formula Weight 155.63
Percent Purity 97%
Assay ee 98%
Chemical Name or Material (R)-(+)-1-(4-Chlorophenyl)ethylamine
Show More Show Less
Hazard Category H302-H315-H317-H318-H330
Hazard Statement GHS H Statement
H301-H330-H314-H318
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only