missing translation for 'onlineSavingsMsg'
Learn More

Quinoxaline-5-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog No. AC429580010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC429580010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC429580010 Supplier Thermo Scientific Chemicals Supplier No. 429580010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6924-66-9 | C9H6N2O2 | 174.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6924-66-9
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.16
MDL Number MFCD02854494
InChI Key QLZNISOPACYKOR-UHFFFAOYSA-N
Synonym 5-quinoxalinecarboxylic acid, 5-carboxyquinoxaline, 5-quinoxalinecarboxylicacid
PubChem CID 776833
IUPAC Name quinoxaline-5-carboxylic acid
SMILES C1=CC(=C2C(=C1)N=CC=N2)C(=O)O

Specifications

Melting Point 167.6°C to 171.7°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 174.16
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Quinoxaline-5-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only