missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinoline 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | -15°C |
| Color | Jaune |
| Boiling Point | 238°C |
| Quantity | 25 g |
| UN Number | 2656 |
| Formula Weight | 129.16 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquide |
| Chemical Name or Material | Quinoline |
Sécurité et manipulation
| EINECSNumber | (5)-0794 |
| RTECSNumber | VA9275000 |
| TSCA | Yes |