missing translation for 'onlineSavingsMsg'
Learn More

Quinoline Ethiodide 98.0+%, TCI America™

Catalog No. E017325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E017325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E017325G Supplier TCI America Supplier No. E017325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 634-35-5
Molecular Formula C11H12IN
Molecular Weight (g/mol) 285.128
MDL Number MFCD00041996
InChI Key PMYUGMDDIBOXQM-UHFFFAOYSA-M
Synonym 1-ethylquinolinium iodide, quinoline ethiodide, 1-ethylquinolin-1-ium iodide, n-ethylquinolinium iodide, quinolinium, 1-ethyl-, iodide, quinolinium, 1-ethyl-, iodide 1:1, 1-ethylquinoliniumiodide, acmc-1b72m, quinoline ethyl iodide
PubChem CID 69446
IUPAC Name 1-ethylquinolin-1-ium;iodide
SMILES CC[N+]1=CC=CC2=CC=CC=C21.[I-]

Specifications

Melting Point 157°C
Color Yellow
Quantity 25 g
Formula Weight 285.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Quinoline Ethiodide
EINECSNumber (5)-0802
RTECSNumber VC3677000
TSCA Yes