missing translation for 'onlineSavingsMsg'
Learn More

Quinoline, 99%

Catalog No. AC221145000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221145000 500 g Glass bottle
AC221140050 5 g Glass bottle
AC221141000 100 g Glass bottle
AC221140025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC221145000 Supplier Thermo Scientific Chemicals Supplier No. 221145000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 91-22-5 | C9H7N | 129.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91-22-5
Molecular Formula C9H7N
Molecular Weight (g/mol) 129.16
MDL Number MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987
InChI Key SMWDFEZZVXVKRB-UHFFFAOYSA-N
Synonym chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol
PubChem CID 7047
ChEBI CHEBI:17362
IUPAC Name quinoline
SMILES C1=CC=C2N=CC=CC2=C1

Specifications

Melting Point -15.0°C
Color Gray to Purple
Density 1.0950g/mL
Boiling Point 237.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6230 to 1.6260
Quantity 500 g
Beilstein 20,339
Fieser 01,975; 02,356; 09,403
Merck Index 15,8185
Specific Gravity 1.095
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in hot water,miscible with alcohol,ether,acetone,benzene,and carbon disulfide
Formula Weight 129.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Quinoline, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing genetic defects.
Harmful in contact with skin.
Toxic
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Call a POISON CENT
EINECSNumber 202-051-6
RTECSNumber VA9275000
TSCA TSCA

RUO – Research Use Only