missing translation for 'onlineSavingsMsg'
Learn More

Quinacridone 93.0+%, TCI America™

Catalog No. Q00575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00575G 5 g
Q005725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00575G Supplier TCI America Supplier No. Q00575G
Only null left

Chemical Identifiers

CAS 1047-16-1
Molecular Formula C20H12N2O2
Molecular Weight (g/mol) 312.33
MDL Number MFCD00059956
InChI Key NRCMAYZCPIVABH-UHFFFAOYSA-N
Synonym 5,12-Dihydroquino[2,3-b]acridine-7,14-dione, Pigment Violet 19
PubChem CID 13976
IUPAC Name 5,7,12,14-tetrahydro-5,12-diazapentacene-7,14-dione
SMILES O=C1C2=CC=CC=C2NC2=CC3=C(NC4=CC=CC=C4C3=O)C=C12

Specifications

Melting Point 390°C
Color Purple-Red
Quantity 5 g
Formula Weight 312.33
Percent Purity ≥93.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Quinacridone
EINECSNumber (5)-1167&(5)-3291
RTECSNumber VA8533500
TSCA Yes