Learn More
Cell Biolabs Inc QuickTiter™ Retrovirus Quantitation Kit, 20 assays
Numéro de catalogue. NC9950016
Encompass
Cell Biolabs' QuickTiter™ Retrovirus Quantitation Kit provides an ultra-fast 45-60 minute procedure that measures physical titer of purified virus or unpurified supernatant via the viral nucleic acid content. Quantitation is performed on a 96-well fluorescence based plate reader. The kit provides sufficient reagents for 20 assays.
Numéro de catalogue. NC9950016 Fournisseur Cell Biolabs Inc Code fournisseur VPK120
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Cell Biolabs' QuickTiter™ Retrovirus Quantitation Kit provides an ultra-fast 45-60 minute procedure that measures physical titer of purified virus or unpurified supernatant via the viral nucleic acid content. Quantitation is performed on a 96-well fluorescence based plate reader. The kit provides sufficient reagents for 20 assays.