Learn More
CBC Tourniquet Quick Release Buckle Tourniquet
Catalog No. 361042428
Change view
Click to view available options
Color:
Light Blue
Catalog No. Color
361042428 Light Blue
1 options
Catalog No. 361042428 Supplier CBC Tourniquet Supplier No. 118790
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CBC quick-release tourniquet stops venous blood flow so that it is easier to locate and puncture a vein.
  • This tourniquet features a solid plastic catch with a plastic buckle that allows for quick release with the touch of a finger.
  • The Quick-Release Buckle Tourniquet is made from heavy-duty woven nylon.
  • Dimensions: 18 x 1 in. (45.7 x 2.5 cm).
  • Latex-free
Specifications
Color Light Blue
Latex Free Yes
Product Type Tourniquet
Disposable Single-use