missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ QuEChERS 500 mg Silica Sorbent with C18 (endcapped) in 50 mL Centrifuge Tubes

Catalog No. 01670173
Change view
Click to view available options
Volume (Metric):
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01670173 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01670173 Supplier Thermo Scientific™ Supplier No. VDQ2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optimize your sample cleanup with Thermo Scientific™ QuEChERS Silica Sorbent with C18 (endcapped) in Centrifuge Tubes. These tubes provide efficient and reliable cleanup for QuEChERS extractions, ensuring high-quality results.

Thermo Scientific QuEChERS Silica Sorbent with C18 (endcapped) in Centrifuge Tubes are designed for efficient sample cleanup in QuEChERS extractions. The pre-measured sorbent ensures consistent performance, while the endcapped C18 provides enhanced retention of non-polar compounds. Packaged in convenient centrifuge tubes, these sorbents streamline your laboratory workflow.

  • Silica sorbent with C18 (endcapped) for effective sample cleanup
  • Centrifuge tubes for convenient handling and processing
  • Enhances retention of non-polar compounds in QuEChERS extractions
  • Pre-measured sorbent ensures consistent and reliable results
  • Streamlines laboratory workflow and reduces preparation time

Specifications

For Use With (Application) QuEChERS
Product Line Quechers
Sorbent Sodium Sulfate
Type QuEChERS Sample Prep
Volume (Metric) 50 mL
Method QuEChERS sample preparation
Sample Size 500 mg per sample
Sample Type All