missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ Flex System Verification Assay Kit

Catalog No. Q33254
Encompass
Change view
Click to view available options
Quantity:
25 assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q33254 25 assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q33254 Supplier Invitrogen™ Supplier No. Q33254

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A fast, easy-to-use, reagent-based assay to test the performance of the Qubit™ Flex Fluorometer.

The Qubit™ Flex System Verification Assay Kit is a fast, easy-to-use, reagent-based assay that tests the performance of the Qubit™ Flex Fluorometer. The kit consists of three components: a blank reagent solution, a green fluorescent reagent, and a far-red fluorescent reagent. The green fluorescent reagent is designed to test blue excitation (470 nm LED; 430-495 nm excitation filter) and green fluorescence detection (510-580 nm emission filter). The far-red fluorescent reagent is designed to test red excitation (635 nm LED; 600-645 nm excitation filter) and far-red fluorescence detection (665-720 nm emission filter).

Note: The Qubit™ Flex System Verification Assay Kit can only be used with the Qubit™ Flex Fluorometer.

Specifications

Product Line Qubit
Type System Verification Assay Kit
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Quantity 25 assays
For Use With (Equipment) Qubit Flex Fluorometer
Does the Qubit Flex NGS Starter Kit (Cat. No. Q45893) include the Qubit Flex System Verification Assay Kit (Cat. No. Q33254)?

No. The Qubit Flex NGS Starter Kit (Cat. No. Q45893) does not include the Qubit Flex System Verification Assay Kit (Cat. No. Q33254). It must be purchased separately.

The description of the Qubit Flex Starter Packages (Cat. Nos. Q32894, Q45893, Q45894) can be found on the Qubit Flex Starter Packages product page.

For Research Use Only. Not for use in diagnostic procedures.