missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ Flex Assay Reservoirs

Catalog No. Q33253
Encompass
Change view
Click to view available options
Quantity:
100 reservoirs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q33253 100 reservoirs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q33253 Supplier Invitrogen™ Supplier No. Q33253

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reservoirs that facilitate the repeated multi-channel of Qubit™ Reagents

Qubit™ Flex Assay Reservoirs facilitate the repeated multi-channel pipetting of Qubit™ reagents into Qubit™ Flex Assay Tube Strips during use of the Qubit™ Flex Fluorometer.

Specifications

Product Line Qubit
Type Assay Reservoir
Shipping Condition Room Temperature
Quantity 100 reservoirs
For Use With (Equipment) Qubit Fluorometer

For Research Use Only. Not for use in diagnostic procedures.