missing translation for 'onlineSavingsMsg'
Learn More

Quartz rod, fused, 2.0mm (0.079in) dia

Catalog No. AA42292NT
Change view
Click to view available options
Quantity:
12 in.
48 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42292NT 48 in.
AA42292NK 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42292NT Supplier Thermo Scientific Chemicals Supplier No. 042292NT

CAS: 14808-60-7 | O2Si | 60.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Boiling Point 2230°C
Melting Point 1710°C
Physical Form Rod
Quantity 48 in.
Packaging Tube
Chemical Name or Material Quartz rod
Hazard Category H350-H373
Precautionary Statement P201-P202-P260-P281-P308+P313-P501c
EINECSNumber 238-878-4
RTECSNumber VV7330000
TSCA U
Recommended Storage Ambient temperatures

RUO – Research Use Only