Learn More
BINDER™ Qualification Documents IQ/OQ
Description
- Parameters depending on unit
- Available as hardcopy inside folder or PDF document
Spécifications
Spécifications
| For Use With (Equipment) | BINDER Chambers Series KBF P, KBF LQC, KBW, KBWF |
| Description | Qualification documents - Digital in PDF format, IQ/OQ documents - supporting documents for validation performed by customers, consisting of: IQ/OQ checklists incl. calibration guide and comprehensive unit documentation; parameters: temperature, humidity, and light values |
| Duration | On request |
| Product Type | IQ/OQ Documentation (PDF) |
| Service Offering | Supporting documents for validation performed by customers |
| Includes | IQ/OQ checklists include calibration guide and comprehensive unit documentation |