missing translation for 'onlineSavingsMsg'
Learn More

Qorpak™ Unlined Polypropylene Caps

Numéro de catalogue. p-4603628
Change view
Click to view available options
Neck Finish:
120-400
33-430
48-400
53-400
89-400
No. per Case:
1300 Caps/Cs.
1700 Caps/Cs.
1800 Caps/Cs.
242 Caps/Cs.
456 Caps/Cs.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Neck Finish No. per Case
02911447 33-430 1800 Caps/Cs.
02991144 48-400 1700 Caps/Cs.
02991145 53-400 1300 Caps/Cs.
02991122 89-400 456 Caps/Cs.
02991109 120-400 242 Caps/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Numéro de catalogue. 02911447 Fournisseur Qorpak™ Code fournisseur 325915
Il en reste null

Polypropylene caps are known for good impact strength, durability, cost-effectiveness, thermal resistance, and pliability.

  • Autoclavable
  • Well-suited for many dry products
  • Add a PTFE disc liner to the unlined polypropylene cap for better leak resistance when autoclaving

Spécifications

Color White
Material Polypropylene
No. per Case 1800 Caps/Cs.
Sterility Non-sterile
Neck Finish 33-430
Disposable Reusable
Product Type Cap
For Use With (Equipment) Wide Mouth Bottle
Diameter (Metric) 33 mm
Autoclavable Autoclavable