missing translation for 'onlineSavingsMsg'
Learn More

Qorpak™ Unlined Round Cans with Triple Tite Lid

Permeation protection against light, oxygen, other gases and essential oils

$321.97 - $4,844.78

Specifications

Product Type Open Top Can
Color Silver
For Use With (Application) For paints, varnishes, pastes and adhesives, protection against light, oxygen, other gases and essential oil
Material Metal
View More Specs

Products 12
Catalog Number Mfr. No. Capacity (English) Includes Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Includes Quantity Price Quantity & Availability  
02991227
View Documents Promotion Details
Qorpak™
MET03094
32 oz. Lid, Securing Clips 24 Cs.
Case of 24 Each for $591.78
Only null left
 
02991229
Accessories View Documents Promotion Details
Qorpak™
MET03090
16 oz. Lid, Securing Clips 24 Cs.
Case of 24 Each for $441.63
Only null left
 
02991234
View Documents Promotion Details
Qorpak™
MET03088
8 oz. Lid, Securing Clips 48 Cs.
Case of 48 Each for $931.81
Only null left
 
02991233
View Documents Promotion Details
Qorpak™
MET03093
32 oz. Lid 56 Cs.
Case of 56 Each for $1,041.18
Only null left
 
02991232
View Documents Promotion Details
Qorpak™
MET03086
4 oz. Lid, Securing Clips 48 Cs.
Case of 48 Each for $1,263.97
Only null left
 
02991235
View Documents Promotion Details
Qorpak™
MET03097
64 oz. Lid, Securing Clips 12 Cs.
Case of 12 Each for $486.13
Only null left
 
02991239
View Documents Promotion Details
Qorpak™
MET03099
128 oz. Lid, Ears, Bails, Securing Clips 6 Cs.
Case of 6 Each for $321.97
Only null left
 
361021201
View Documents Promotion Details
Qorpak™
MET03098
128 oz. Triple Tite Lid 46 Cs.
Case of 34 Each for $1,108.47
Only null left
 
02991249
View Documents Promotion Details
Qorpak™
ALP MET03101
128 oz. Lid, Ears, Bails, Securing Clips 46 Cs.
Case of 34 Each for $1,340.94
Only null left
 
02991280
View Documents Promotion Details
Qorpak™
MET03087
8 oz. Lid 340 Cs.
Case of 340 Each for $4,844.78
Only null left
 
02991281
View Documents Promotion Details
Qorpak™
MET03096
64 oz. Lid 75 Cs.
Case of 75 Each for $2,517.47
Only null left
 
02991282
View Documents Promotion Details
Qorpak™
MET03089
16 oz. Lid 52 Cs.
Case of 52 Each for $892.59
Only null left
 
Description

Description

  • Features resealable multi-friction plug lid for more than one point of contact between lid and can
  • Provides an airtight seal when opened by prying and sealed by pressing
  • May be sterilized by heat
  • Used for paints, varnishes, pastes and adhesives
Specifications

Specifications

Open Top Can
Silver
For paints, varnishes, pastes and adhesives, protection against light, oxygen, other gases and essential oil
Metal
Videos
SDS
Documents

Documents