Learn More
Qorpak™ Unlined Round Cans with Triple Tite Lid
Permeation protection against light, oxygen, other gases and essential oils
Supplier:  Qorpak™ MET03093
 View more versions of this product
Catalog No. 02991233
Only null left
Description
Description
  • Features resealable multi-friction plug lid for more than one point of contact between lid and can
  • Provides an airtight seal when opened by prying and sealed by pressing
  • May be sterilized by heat
  • Used for paints, varnishes, pastes and adhesives
Specifications
Chemical Identifiers
Open Top Can
Silver
For paints, varnishes, pastes and adhesives, protection against light, oxygen, other gases and essential oil
Metal
Specifications
Open Top Can
946.3 mL
For paints, varnishes, pastes and adhesives, protection against light, oxygen, other gases and essential oil
Lid
4.875 in.
4.25 in.
32 oz.
Silver
Metal
56 Cs.
414 mm
Product Suggestions
Product Suggestions