Learn More
Qorpak™ CurTec™ Lids for White Packo Containers
For use with Packo containers. Qorpak™ CurTec™ Lids provides high level of product protection and added value through quality, functionality, ease of use, and design.
Supplier: Qorpak™ 248750
Description
- Made of polypropylene
- Available with and without gasket
Identifiants chimiques
| Polypropylene | |
| Convenience | |
| Reusable |
| 30/Cs. | |
| Non-sterile | |
| Not Autoclavable |
Spécifications
| Red | |
| Polypropylene | |
| Convenience | |
| Reusable | |
| 300 to 1300 mL Packo Containers |
| Gasket | |
| 30/Cs. | |
| Non-sterile | |
| Quarter-turn Screw Lid with Gasket | |
| Not Autoclavable |