Learn More
New England Biolab Ltd Q5 High-Fidelity DNA Polymerase 100 units DFS Item
Supplier:  New England Biolab Ltd M0491S
Q5 High-Fidelity DNA Polymerase is a high-fidelity, thermostable DNA polymerase with 3' to 5' exonuclease activity, fused to a processivity-enhancing Sso7d domain to support robust DNA amplification. With an error rate ~280-fold lower than that of Taq DNA Polymerase, Q5 High-Fidelity DNA Polymerase is ideal for cloning and can be used for long or difficult amplicons. Q5 High-Fidelity DNA Polymerase is supplied with an optimized buffer system that allows robust amplification regardless of GC content. The 5X Q5 Reaction Buffer contains 2 mM Mg++ at final (1X) reaction concentrations and is recommended for most routine applications. For GC-rich targets (>= 65% GC), amplification can be improved by the addition of the 5X Q5 High GC Enhancer. Q5 High-Fidelity DNA Polymerase is unlike typical, lower fidelity PCR enzymes.
Catalog No. NC0355743
Only null left
Explore available promotions