Learn More
Restek™ Q-Sep QuEChERS Extraction Salts
Numéro de catalogue. 06611949
Encompass
Change view
Click to view available options
Material:
4 g MgSO4, 1 g NaCl, 1 g TSCD, 0.5 g DHS
Numéro de catalogue. Material
06611949 4 g MgSO4, 1 g NaCl, 1 g TSCD, 0.5 g DHS
1 options
Numéro de catalogue. 06611949 Fournisseur Restek™ Code fournisseur 25849
Il en reste null
Pre-weighed, ultra-pure extraction salts
  • Easy-open packets eliminate need for a second empty tube for salt transfer
  • Free-flowing salts transfer easily and completely
  • Convenient slim packets fit perfectly into tubes to prevent spills
  • Ready-to-use tubes, no glassware required
  • Ideal for original unbuffered, AOAC (2007.01), and European (EN 15662) QuEChERS methods
Spécifications
Type Q-Sep QuEChERS Extraction Salts
Includes Salt packets
Material 4 g MgSO4, 1 g NaCl, 1 g TSCD, 0.5 g DHS
Quantity 50 packets