missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic Acid Sodium Salt (Reagent), Fisher Chemical

Catalog No. O4376100
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
O4376100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O4376100 Supplier Thermo Fisher Scientific Supplier No. O4376 100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 113-24-6
Molecular Formula C3H3NaO3
Molecular Weight (g/mol) 110.044
MDL Number MFCD00002586
InChI Key DAEPDZWVDSPTHF-UHFFFAOYSA-M
Synonym sodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08
PubChem CID 23662274
ChEBI CHEBI:50144
IUPAC Name sodium;2-oxopropanoate
SMILES CC(=O)C(=O)[O-].[Na+]

Specifications

Melting Point 300°C
Color White
pH 5.5 to 6.5
Assay Percent Range ≥97 %
Packaging Glass Bottle
Linear Formula CH3COCOONa
Quantity 100 g
Formula Weight 110.04
Percent Purity ≥97%
Grade Reagent
Physical Form Powder or Solid
Chemical Name or Material Pyruvic Acid Sodium Salt
Show More Show Less