missing translation for 'onlineSavingsMsg'
Learn More

Pyrromethene 597 98.0+%, TCI America™

Catalog No. B4400200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4400200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4400200MG Supplier TCI America Supplier No. B4400200MG
Only null left

Chemical Identifiers

CAS 137829-79-9
Molecular Formula C22H33BF2N2
Molecular Weight (g/mol) 374.327
MDL Number MFCD09753047
InChI Key SEHGNHOGQDPQRC-UHFFFAOYSA-N
Synonym Pyrromethene 597
PubChem CID 15294197
SMILES [B-]1(N2C(=C(C(=C2C(=C3[N+]1=C(C(=C3C)C(C)(C)C)C)C)C)C(C)(C)C)C)(F)F

Specifications

Melting Point 249°C
Quantity 200 mg
Formula Weight 374.33
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material [[(4-tert-Butyl-3,5-dimethyl-1H-pyrrol-2-yl)(4-tert-butyl-3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]me