missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pyrroloquinoline quinone

Catalog No. p-7214709
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467010010 1 g
AC467012500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467010010 Supplier Thermo Scientific Chemicals Supplier No. 467010010
Only null left

CAS: 72909-34-3 | C14H6N2O8, C14H6N2O8 | 330.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 72909-34-3
Molecular Formula C14H6N2O8

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: slightly soluble.
Formula Weight 330.21
InChI Key MMXZSJMASHPLLR-UHFFFAOYSA-N
SMILES OC(=O)C1=CC2=C(N1)C1=C(C=C(N=C1C(=O)C2=O)C(O)=O)C(O)=O
IUPAC Name 4,5-dioxo-1H,4H,5H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid
Molecular Weight (g/mol) 330.21
Chemical Name or Material Pyrroloquinoline quinone
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only