accessibility menu, dialog, popup

UserName

OHOHNOONOHOHOO

CAS RN 72909-34-3

IUPAC Name: 4,5-dioxo-1H,4H,5H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid
Synonyms: pyrroloquinoline quinone pyrroloquinoline-quinone pyrrolo quinoline quinone pyrrolo-quinoline quinone pqq cofactor coenzyme pqq methoxatin coenzyme, pqq cofactor, pqq pqq, coenzyme
Molecular Weight (g/mol): 330.21
Molecular Formula: C14H6N2O8
InChi Key: MMXZSJMASHPLLR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=C(N1)C1=C(C=C(N=C1C(=O)C2=O)C(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)