missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-3-carboxylic Acid 95.0+%, TCI America™

Catalog No. P18991G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P18991G Supplier TCI America Supplier No. P18991G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 931-03-3
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
MDL Number MFCD00800594
InChI Key DOYOPBSXEIZLRE-UHFFFAOYSA-N
Synonym pyrrole-3-carboxylic acid, pyrrole-3-carboxylicacid, 3-carboxy-1h-pyrrole, 1h-pyrrole-3-carboxylicacid, pubchem8345, 3-carboxypyrrole, 3-pyrrolecarboxylic acid, acmc-1agqb, 3-pyrrole-carboxylic acid, pyrrole 3-carboxylic acid
PubChem CID 101030
ChEBI CHEBI:68076
IUPAC Name 1H-pyrrole-3-carboxylic acid
SMILES C1=CNC=C1C(=O)O

Specifications

Melting Point 148°C
Quantity 1 g
Formula Weight 111.10
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyrrole-3-carboxylic Acid