missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUJIFILM Wako PYROSTAR™ ES-F 5.2 mL Bulk Kit
Endotoxin-specific and specially formulated to be unreactive to (1,3)-ß-D-glucan, dual-purpose, formulated to be used as gel clot or kinetic turbidimetric assay, bulk configuration for large volume labs
$28,050.68
| Catalog Number | Mfr. No. | Sensitivity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Sensitivity | Price | Quantity & Availability | |||||
|
12333019
|
FUJIFILM Wako
WPEM50015 |
0.015 EU/mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
12333020
|
FUJIFILM Wako
WPEM50003 |
0.03 EU/mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
12333021
|
FUJIFILM Wako
WPEM50006 |
0.06 EU/mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
12333022
|
FUJIFILM Wako
WPEM50125 |
0.125 EU/mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
12333023
|
FUJIFILM Wako
WPEM50025 |
0.25 EU/mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Endotoxin-specific reagent avoids false positive results from glucans
- Dual-purpose reagent can perform either gel clot or kinetic turbidimetric assay (KTA)
- Kinetic turbidimetric sensitivities range from 0.001 to 10 EU/mL
- Chances of interference reduced by high sensitivity
- Facilitates easier transition from gel clot to KTA
Specifications
| Lyophilized | |
| 5.2 mL | |
| 100 Vials |