missing translation for 'onlineSavingsMsg'
Learn More

FUJIFILM Wako PYROSTAR™ ES-F 5.2 mL Bulk Kit

Catalog No. 12333019
Change view
Click to view available options
Sensitivity:
0.015 EU/mL
0.03 EU/mL
0.06 EU/mL
0.125 EU/mL
0.25 EU/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sensitivity
12333019 0.015 EU/mL
12333020 0.03 EU/mL
12333021 0.06 EU/mL
12333022 0.125 EU/mL
12333023 0.25 EU/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 12333019 Supplier FUJIFILM Wako Supplier No. WPEM50015

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Endotoxin-specific and specially formulated to be unreactive to (1,3)-ß-D-glucan, dual-purpose, formulated to be used as gel clot or kinetic turbidimetric assay, bulk configuration for large volume labs

  • Endotoxin-specific reagent avoids false positive results from glucans
  • Dual-purpose reagent can perform either gel clot or kinetic turbidimetric assay (KTA)
  • Kinetic turbidimetric sensitivities range from 0.001 to 10 EU/mL
  • Chances of interference reduced by high sensitivity
  • Facilitates easier transition from gel clot to KTA

Specifications

Form Lyophilized
Fill Volume 5.2 mL
Includes 100 x 5.2 mL vials of lysate
Quantity 100 Vials
Sensitivity 0.015 EU/mL