Learn More
FUJIFILM Wako PYROSTAR™ ES-F 5.2 mL Bulk Kit
Endotoxin-specific and specially formulated to be unreactive to (1,3)-ß-D-glucan, dual-purpose, formulated to be used as gel clot or kinetic turbidimetric assay, bulk configuration for large volume labs
Supplier: FUJIFILM Wako WPEM50006
Description
- Endotoxin-specific reagent avoids false positive results from glucans
- Dual-purpose reagent can perform either gel clot or kinetic turbidimetric assay (KTA)
- Kinetic turbidimetric sensitivities range from 0.001 to 10 EU/mL
- Chances of interference reduced by high sensitivity
- Facilitates easier transition from gel clot to KTA
Chemical Identifiers
| Lyophilized | |
| 5.2 mL | |
| 100 Vials |
Specifications
| Lyophilized | |
| 100 x 5.2 mL of lysate | |
| 0.06 EU/mL |
| 5.2 mL | |
| 100 Vials |