missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyronin Y
Description
Pyronin Y is a cationic dye that intercalates RNA and is visible as a red band during electrophoresis. It has also been shown to accumulate in mitochondria of viable cells. In conjuction with Methyl Green or Nancy-520, Pyronin Y can be used as a fluorescent differential stain of nucleic acids in paraffin tissue sections.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pyronin Y is a cationic dye that intercalates RNA and is visible as a red band during electrophoresis. It has also been shown to accumulate in mitochondria of viable cells. In conjuction with Methyl Green or Nancy-520, Pyronin Y can be used as a fluorescent differential stain of nucleic acids in paraffin tissue sections.
Solubility
Soluble in water. Also soluble in methyl cellosolve (9 mg/mL) and ethanol (3 mg/mL).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Pyronin Y is a cationic dye that intercalates RNA and is visible as a red band during electrophoresis. It has also been shown to accumulate in mitochondria of viable cells. In conjuction with Methyl Green or Nancy-520, Pyronin Y can be used as a fluorescent differential stain of nucleic acids in paraffin tissue sections.
Solubility
Soluble in water. Also soluble in methyl cellosolve (9 mg/mL) and ethanol (3 mg/mL).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 250°C to 260°C |
| Beilstein | 3795789 |
| Merck Index | 14,8007 |
| Color | Green |
| Solubility Information | Soluble in water. Also soluble in methyl cellosolve (9mg/mL) and ethanol (3mg/mL). |
| Quantity | 5 g |
| Physical Form | Crystalline Powder |
| Formula Weight | 302.8 |
| Chemical Name or Material | Pyronin Y |
Safety and Handling
| Hazard Category | H341 |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| EINECSNumber | 202-147-8 |
| RTECSNumber | BQ1450000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only