missing translation for 'onlineSavingsMsg'
Learn More

Pyrocatechol Violet Indicator, For Metal Titration, Solstice

Catalog No. p-7120744
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046149 1 g Glass Bottle
60046150 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046149 Supplier Solstice Supplier No. 326721G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Indicator for metal titration

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 115-41-3
Molecular Formula C19H14O7S
Molecular Weight (g/mol) 386.37
MDL Number MFCD00005868
InChI Key RRRCKIRSVQAAAS-UHFFFAOYSA-N
Synonym Catechol violet
PubChem CID 66993
IUPAC Name 4-[3-(3,4-dihydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]benzene-1,2-diol
SMILES OC1=CC=C(C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(O)=C1

Specifications

Melting Point 185°C
Packaging Glass Bottle
Suitability For metal titration
Visual Transition Interval 6.5 to 8
UN Number NONH for all modes of transport
Beilstein 353938
Quantity 1 g
Physical Form Powder
Formula Weight 386.38g/mol
Grade Indicator
Chemical Name or Material Pyrocatechol Violet
Show More Show Less
EINECSNumber 204-088-3