missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-5-carboxylic acid, 95%

Catalog No. p-7053752
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61406MD 250 mg
AAH6140603 1 g
AAH6140606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH61406MD Supplier Thermo Scientific Chemicals Supplier No. H61406MD

CAS: 4595-61-3 | C5H4N2O2 | 124.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4595-61-3
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.10
MDL Number MFCD00856162
InChI Key IIVUJUOJERNGQX-UHFFFAOYSA-N
Synonym 5-pyrimidinecarboxylic acid, 5-carboxypyrimidine, pyrimidine-5-carboxilic acid, 5-pyrimidine carboxylic acid, pyrimidine-5-carboxylate acid, pyrimidine-5-carboxylicacid, pubchem2056, acmc-209k5a, 5-pyrimidyl carboxylic acid, pyrimidin-5-carboxylic acid
PubChem CID 78346
IUPAC Name pyrimidine-5-carboxylic acid
SMILES OC(=O)C1=CN=CN=C1

Specifications

Melting Point 259°C to 261°C
Quantity 250 mg
Formula Weight 124.1
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Pyrimidine-5-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-994-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only