missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridoxine Dicaprylate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 72°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 421.58 |
| Percent Purity | ≥98.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Pyridoxine Dicaprylate |
Safety and Handling
| EINECSNumber | (9)-2209 |
| TSCA | No |