missing translation for 'onlineSavingsMsg'
Learn More

Pyridine - Sulfur Trioxide Complex 95.0+%, TCI America™

Catalog No. P0998500G
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0998500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0998500G Supplier TCI America Supplier No. P0998500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26412-87-3
Molecular Formula C5H5NO3S
Molecular Weight (g/mol) 159.16
MDL Number MFCD00012437
InChI Key UDYFLDICVHJSOY-UHFFFAOYSA-N
Synonym pyridine sulfur trioxide, sulfur trioxide pyridine complex, pyridine; sulfur trioxide, pyridine sulfotrioxide, pyridine compound with sulfur trioxide 1:1, sulfur trioxide, compd. with pyridine, pyridine-sulfur trioxide complex, active sulfure trioxide, sulfur trioxide-pyridine, pyridine-sulfur trioxide
PubChem CID 168533
IUPAC Name pyridine; sulfonylideneoxidane
SMILES O=S(=O)=O.C1=CC=NC=C1

Specifications

Color White-Yellow
Quantity 500 g
UN Number 3261
Formula Weight 159.16
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridine - Sulfur Trioxide Complex