missing translation for 'onlineSavingsMsg'
Learn More

Pyridine Hydrobromide 98.0+%, TCI America™

Catalog No. P166625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P166625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P166625G Supplier TCI America Supplier No. P166625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18820-82-1
Molecular Formula C5H6BrN
Molecular Weight (g/mol) 160.014
MDL Number MFCD00012801
InChI Key BBFCIBZLAVOLCF-UHFFFAOYSA-N
Synonym pyridine hydrobromide, pyridinium bromide, pyridine, hydrobromide, pyridine, hydrobromide 1:1, pyridine hbr, pyridinehydrobromide, acmc-1bovh, ksc178o2t, pyridine hydrobromide, purum at
PubChem CID 87810
IUPAC Name pyridine;hydrobromide
SMILES C1=CC=NC=C1.Br

Specifications

Melting Point 217°C
Quantity 25 g
Formula Weight 160.01
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridine Hydrobromide