missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, CHROMASOLV™ Plus, for HPLC, ≥99.9%, Solstice

Catalog No. 6001314
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001314 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001314 Supplier Solstice Supplier No. 2704071L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHROMASOLV™ Plus, for HPLC,≥99.9%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Melting Point -42°C (literature)
Absorbance max. 0.01 at 400 nm, max. 0.01 at 350 nm, max. 0.02 at 335 nm, max. 0.15 at 315 nm, max. 1.0 at 305 nm
CAS Max % 100.0000%
Density 0.978 g/mL at 25 °C(lit.)
Quantity 1 L
Flash Point 20°C
Packaging Glass Bottle
Linear Formula C5H5N
UN Number UN1282
Beilstein 103233
Boiling Point 115°C
Formula Weight 79.1
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Pyridine, CHROMASOLV™ Plus, for HPLC, ≥99.9%
Show More Show Less
ShelfLife 450 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Pyridine
EINECSNumber 203-809-9