missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine-Barbituric Acid Reagent, for Cyanide Determination, Ricca Chemical
For Cyanide Determination ASTM, APHA
$389.37 - $840.47
Specifications
| Chemical Name or Material | Pyridine-Barbituric Acid Reagent |
|---|---|
| Color | Undesignated |
| CAS | 7732-18-5 , 110-86-1 , 7647-01-0 , 67-52-7 |
| CAS Min % | 69.0,20.27,4.87,4.13 |
| CAS Max % | 72.0,21.1,5.07,4.3 |