missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, B&J Brand™, for GC and non-aqueous titrations, >99.8%, Honeywell Burdick & Jackson

Catalog No. 6001316
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001316 6 x 1 L Glass Bottle
6001315 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001316 Supplier Honeywell Chemicals Supplier No. 3321L
Only null left

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

CAS Max % 100.0000%
Density 0.983 g/cm3
Quantity 6 x 1 L
Flash Point 20°C
Packaging Glass Bottle
UN Number UN1282
Beilstein 103233
Boiling Point 115.25°C
Formula Weight 79.1
Percent Purity >99.8%
Grade aqueous titrations
Chemical Name or Material Pyridine, B&J Brand™, for GC and non-aqueous titrations, >99.8%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Pyridine