missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, 99+%, ACS reagent

Numéro de catalogue. AC418540250
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Metal Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC418540250 25 L Metal Drum
AC418545000 500 mL EcoSafPak™ Glass Bottle
AC418540010 1 L Glass Bottle
AC418540025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. AC418540250 Fournisseur Thermo Scientific Chemicals Code fournisseur 418540250
Il en reste null

CAS: 110-86-1 | C5H5N | 79.102 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Spécifications

Melting Point -42.0°C
Color Colorless
Density 0.9780g/mL
Quantity 25 L
Flash Point 17°C
Packaging Metal Drum
Physical Form Liquid
Refractive Index 1.5085 to 1.5105
Residue after Evaporation 0.002% max.
Specific Gravity 0.978
Assay Percent Range 99+%
Vapor Pressure Vapor Pressure: 20mbar at 20°C
Viscosity 0.95 mPa.s (20°C)
Water 0.1% max. (K.F.)
UN Number 1282
Beilstein 20, 181
Fieser 01,958; 02,349; 04,414; 06,497; 11,448; 12,416; 13,293; 15,1; 17,153
Merck Index 15, 8081
Boiling Point 115.0°C to 116.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and most other common, organic solvents
Formula Weight 79.1
Percent Purity ≥99%
Grade ACS
Chemical Name or Material Pyridine
Afficher plus de résultats Afficher moins de résultats
Hazard Category Danger
Hazard Statement GHS P Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-809-9
RTECSNumber UR8400000
TSCA TSCA

RUO – Research Use Only