missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine, 99.8%, for biochemistry, AcroSeal™, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -42.0°C |
| Density | 0.9780g/mL |
| Boiling Point | 115.0°C to 116.0°C |
| Flash Point | 17°C |
| Assay Percent Range | 99.75% min. (GC) |
| Packaging | AcroSeal™ Glass bottle |
| Refractive Index | 1.5085 to 1.5105 |
| Quantity | 1 L |
| Specific Gravity | 0.978 |
| Merck Index | 15,8081 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H StatementHarmful in contact with skin.Harmful if inhaled.Harmful if swallowed.Causes skin irritation.Causes serious eye irritation.Highly flammable liquid and vapor. |
| Precautionary Statement | GHS P StatementWear protective gloves/protective clothing/eye protection/face protection.IF ON SKIN: Wash with plenty of soap and water.IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. |
| EINECSNumber | 203-809-9 |
RUO – Research Use Only