missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-4-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog No. AAL1785403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1785403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1785403 Supplier Thermo Scientific Chemicals Supplier No. L1785403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 181219-01-2 | Molecular Formula: C11H16BNO2 | Molecular Weight: 205.06 g/mol

It is employed in the synthesis of new pyridazino[4,5-b]indol-4-ones and pyridazin-3(2H)-one analogs as DYRK1A inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of new pyridazino[4,5-b]indol-4-ones and pyridazin-3(2H)-one analogs as DYRK1A inhibitors.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 181219-01-2
Molecular Formula C11H16BNO2
Molecular Weight (g/mol) 205.06
MDL Number MFCD01319051
InChI Key NLTIETZTDSJANS-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 4-pyridineboronic acid pinacol ester, pyridine-4-boronic acid pinacol ester, 4-pyridylboronic acid pinacol ester, pyridine-4-boronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine-4-boronic pinacolate, pyridine-4-boronic acid pinacol cyclic ester, 2-4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pyridine-4-boronic pinacol ester
PubChem CID 2734648
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CC=NC=C1

Specifications

Melting Point 150°C to 152°C
Quantity 1 g
Beilstein 7919059
Solubility Information Insoluble in water.
Formula Weight 205.07
Percent Purity 98%
Chemical Name or Material Pyridine-4-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only