missing translation for 'onlineSavingsMsg'
Learn More

pyridine-3-sulfonyl chloride hydrochloride, 95%, Thermo Scientific™

Catalog No. CC04103DA
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC04103DA 1 g
CC04103CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC04103DA Supplier Thermo Scientific Chemicals Supplier No. CC04103DA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 42899-76-3
Molecular Formula C5H5Cl2NO2S
Molecular Weight (g/mol) 214.06
InChI Key VIVPWOMJFLICOZ-UHFFFAOYSA-N
Synonym pyridine-3-sulfonyl chloride hydrochloride, pyridine-3-sulfonyl chloride hcl, 3-pyridinesulfonyl chloride hydrochloride, pyridine-3-sulphonyl chloride hydrochloride, 3-pyridinesulfonyl chloride, hcl, 3-pyridinesulfonyl chloride, hydrochloride, 3-pyridylsulfonyl chloride hydrochloride, pyridine-3-sulfonylchloride hydrochloride, pyridin-3-ylsulfonyl chloride hydrochloride, 3-pyridylsulfonylchloride hcl
PubChem CID 12571517
IUPAC Name pyridine-3-sulfonyl chloride;hydrochloride
SMILES C1=CC(=CN=C1)S(=O)(=O)Cl.Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 214.07
Percent Purity 95%
Chemical Name or Material Pyridine-3-sulfonyl chloride hydrochloride
Hazard Category Danger