missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3-sulfonyl chloride hydrochloride, 95%, Thermo Scientific Chemicals

Catalog No. p-7077364
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451480050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC451480050 Supplier Thermo Scientific Chemicals Supplier No. 451480050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42899-76-3
Molecular Formula C5H4ClNO2S·ClH
Molecular Weight (g/mol) 214.07
InChI Key VIVPWOMJFLICOZ-UHFFFAOYSA-N
Synonym pyridine-3-sulfonyl chloride hydrochloride, pyridine-3-sulfonyl chloride hcl, 3-pyridinesulfonyl chloride hydrochloride, pyridine-3-sulphonyl chloride hydrochloride, 3-pyridinesulfonyl chloride, hcl, 3-pyridinesulfonyl chloride, hydrochloride, 3-pyridylsulfonyl chloride hydrochloride, pyridine-3-sulfonylchloride hydrochloride, pyridin-3-ylsulfonyl chloride hydrochloride, 3-pyridylsulfonylchloride hcl
PubChem CID 12571517
IUPAC Name pyridine-3-sulfonyl chloride;hydrochloride
SMILES C1=CC(=CN=C1)S(=O)(=O)Cl.Cl

Specifications

Melting Point 144.0°C to 145.0°C
Elemental Analysis 26.37 to 29.74% (C) 2.21 to 2.50% (H) 6.15 to 6.94% (N)
Infrared Spectrum Authentic
Assay Percent Range 94% min. as diethylsulfonamide (HPLC)
Quantity 5 g
Formula Weight 214.07
Percent Purity 95%
Chemical Name or Material Pyridine-3-sulfonyl chloride hydrochloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair

RUO – Research Use Only