missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3,4-dicarboxylic acid, 98%

Catalog No. AAA1458018
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1458018 50 g
AAA1458009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1458018 Supplier Thermo Scientific Chemicals Supplier No. A1458018

CAS: 490-11-9 | C7H5NO4 | 167.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 490-11-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00006392
InChI Key MUYSADWCWFFZKR-UHFFFAOYSA-N
Synonym 3,4-pyridinedicarboxylic acid, cinchomeronic acid, chinchomeronic acid, 3,4-pyridinedicarboxylicacid, pyridine-3,4-dicarboxylicacid, unii-7389uzw5dy, 3,4-pyridindicarbons, cinchomeronsaure, pubchem2419, 3,4-dicarboxypyridine
PubChem CID 10273
ChEBI CHEBI:46860
IUPAC Name pyridine-3,4-dicarboxylic acid
SMILES OC(=O)C1=CC=NC=C1C(O)=O

Specifications

Melting Point ∼255°C (decomposition)
Quantity 50 g
Beilstein 137242
Merck Index 14,2283
Formula Weight 167.12
Percent Purity 98%
Chemical Name or Material Pyridine-3,4-dicarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-705-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only