missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2-carboxylic Acid 99.0+%, TCI America™

Catalog No. P0421500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0421500G 500 g
P042125G 25 g
P0421100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0421500G Supplier TCI America Supplier No. P0421500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98-98-6 | C6H5NO2 | 123.111 g/mol

Chemical Identifiers

CAS 98-98-6
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.111
MDL Number MFCD00006293
InChI Key SIOXPEMLGUPBBT-UHFFFAOYSA-N
Synonym picolinic acid, 2-pyridinecarboxylic acid, 2-picolinic acid, 2-carboxypyridine, alpha-picolinic acid, o-pyridinecarboxylic acid, pyridinecarboxylic acid, alpha-pyridinecarboxylic acid, picolinate, acide picolique
PubChem CID 1018
ChEBI CHEBI:28747
IUPAC Name pyridine-2-carboxylic acid
SMILES C1=CC=NC(=C1)C(=O)O

Specifications

Melting Point 140°C
Color White
Quantity 500 g
Formula Weight 123.11
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridine-2-carboxylic Acid
EINECSNumber (9)-1077
RTECSNumber TJ7344000
TSCA Yes