missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ VISTA™ Test Tubes with Beaded Rim

Functionality, economy, and quality.

Supplier:  Corning™ 7080020/EM

 Afficher plus de versions de ce produit

Numéro de catalogue. S76110L


Il en reste null

Description

Description

  • Well-annealed
  • Fire-polished rims
  • Heat-resistant and chemically stable
  • With blue marking spot (except 0.98 in. [25mm] O.D. tubes)

Spécifications

Identifiants chimiques

Beaded Rim
Marking Spot
No
Borosilicate Glass
Non-sterile
Not Included

Spécifications

Beaded Rim
150 mm
Marking Spot
No
Not Included
34 mL
Borosilicate Glass
Non-sterile
20 mm
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions