Learn More
Corning™ PYREX™ Thermometer Adapter
GREENER_CHOICE
Component part for organic chemistry kit
$60.02 - $161.11
Spécifications
For Use With (Application) Replacement rubber thermometer holders
Includes Rubber thermometer holder
Material Borosilicate Glass
Product Type Straight Adapter
For Use With (Equipment) Thermometers
voir les spécifications
Produits 3
Numéro de catalogue Numéro du manufacturier. Length (Metric) Length (English) Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Length (Metric) Length (English) Prix Quantity & Availability  
010341A
GREENER_CHOICE
Afficher les documents Détails de la promotion
Corning™
882114
60 mm 2.36 in.
caisse for $60.02
Il en reste null
010341B
GREENER_CHOICE
View Documents Promotion Details
Corning™
882119
57 mm 2.24 in.
caisse for $104.20
Only null left
010341C
GREENER_CHOICE
View Documents Promotion Details
Corning™
882124
80 mm 3.14 in.
caisse for $161.11
Only null left
Description
Description
One end with a standard taper ground joint and the other end tooled to accommodate a thermometer. The adapter is supplied complete with a rubber thermometer holder (Cat. No. 7715).
This tube is also a replacement part for all organic chemistry kit (Cat. No. 6949 through 6949K).
Specifications
Specifications
Replacement rubber thermometer holders
Borosilicate Glass
Thermometers
Rubber thermometer holder
Straight Adapter