Learn More
Corning™ PYREX™ Accelerated One-Step™ Extractor/Concentrator Replacement Parts
Catalog No. 0777084
Change view
Click to view available options
Product Type:
Extractor Body
Photoceram Membrane Support
PTFE Membrane (47 mm)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
0777084 Photoceram Membrane Support
0777079 Extractor Body
0777083 PTFE Membrane (47 mm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0777084 Supplier Corning™ Supplier No. 391847MS
Get Up to $2,500 in Products with Purchase
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with Accelerated One-Step Extractor/Concentrator Systems
Replacement part for Pyrex Accelerated One-Step extractor/concentrator system. The system performs extraction, concentration, and drying with the same apparatus and is designed for use with solvents of heavier specific gravity than sample being extracted.
Specifications
Product Type Photoceram Membrane Support
For Use With (Equipment) Pyrex Accelerated One-Step Extractor or Concentrator System
Length (English) 1.85 in.
Length (Metric) 4.7 cm