missing translation for 'onlineSavingsMsg'
Learn More

Pyrene, 98%

Catalog No. AAL0816214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0816214 25 g
AAL0816222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0816214 Supplier Thermo Scientific Chemicals Supplier No. L0816214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 129-00-0 | C16H10 | 202.256 g/mol

Pyrene and its derivatives are used commercially to make dyes and dye precursors, for example pyranine and naphthalene-1,4,5,8-tetracarboxylic acid. It is used as a probe to determine solvent environments and fluorescence spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrene and its derivatives are used commercially to make dyes and dye precursors, for example pyranine and naphthalene-1,4,5,8-tetracarboxylic acid. It is used as a probe to determine solvent environments and fluorescence spectroscopy.

Solubility
Soluble in toluene. Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Chemical Identifiers

CAS 129-00-0
Molecular Formula C16H10
Molecular Weight (g/mol) 202.256
MDL Number MFCD00004136
InChI Key BBEAQIROQSPTKN-UHFFFAOYSA-N
Synonym benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene
PubChem CID 31423
ChEBI CHEBI:39106
IUPAC Name pyrene
SMILES C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2

Specifications

Melting Point 148°C to 152°C
Density 1.271
Boiling Point 393°C
Flash Point 208°C (406°F)
Quantity 25 g
Beilstein 1307225
Merck Index 14,7963
Solubility Information Soluble in toluene. Insoluble in water.
Formula Weight 202.26
Percent Purity 98%
Chemical Name or Material Pyrene
Show More Show Less
Hazard Category H336-H373
Hazard Statement GHS H Statement
H332
Harmful if inhaled.
Precautionary Statement P260-P271-P304+P340-P312-P314-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 204-927-3
RTECSNumber UR2450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only