missing translation for 'onlineSavingsMsg'
Learn More

Pyrazolo[1,5-a]pyridine-7-carboxaldehyde, 97%

Catalog No. AAH50074MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50074MD 250 mg
AAH5007403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50074MD Supplier Thermo Scientific Chemicals Supplier No. H50074MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 362661-83-4 | C8H6N2O | 146.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 362661-83-4
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.149
MDL Number MFCD06739029
InChI Key SGNBVQKSJWCGOJ-UHFFFAOYSA-N
Synonym pyrazolo 1,5-a pyridine-7-carbaldehyde, pyrazolo 1,5-a pyridine-7-carboxaldehyde, 8-hydropyrazolo 1,5-a pyridine-7-carbaldehyde, pyrazolo 1,5-a pyridine-7-carboxaldehyde 9ci, pyrazolo 1,5-alpha pyridine-7-carboxaldehyde
PubChem CID 10866474
IUPAC Name pyrazolo[1,5-a]pyridine-7-carbaldehyde
SMILES C1=CC2=CC=NN2C(=C1)C=O

Specifications

Melting Point 83°C to 86°C
Quantity 250 mg
Formula Weight 146.15
Percent Purity 97%
Chemical Name or Material Pyrazolo[1,5-a]pyridine-7-carboxaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only