missing translation for 'onlineSavingsMsg'
Learn More

Pyrazine-2-carbonyl chloride, 90+%, Thermo Scientific Chemicals

Catalog No. AAH5041803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5041803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5041803 Supplier Thermo Scientific Chemicals Supplier No. H5041803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19847-10-0 | Molecular Formula: C5H3ClN2O | Molecular Weight: 142.542 g/mol

Pyrazine-2-carbonyl chloride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrazine-2-carbonyl chloride is used as pharmaceutical intermediates.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas. Incompatible with bases, oxidizing agents and moisture.

Chemical Identifiers

CAS 19847-10-0
Molecular Formula C5H3ClN2O
Molecular Weight (g/mol) 142.542
MDL Number MFCD00792886
InChI Key TXJKATOSKLUITR-UHFFFAOYSA-N
Synonym 2-pyrazinecarbonyl chloride, pyrazinecarbonyl chloride, pyrazine-2-carboxylic acid chloride, 2-pyrazinecarbonylchloride, pyrazine 2-carbonyl chloride, 2-pyrazine carbonyl chloride, pyrazinoyl chloride, acmc-20ann0, 2-pyrazine carboxylic acid chloride, pyrazine-2-carbonyl chloride, technical 1g
PubChem CID 88279
IUPAC Name pyrazine-2-carbonyl chloride
SMILES C1=CN=C(C=N1)C(=O)Cl

Specifications

Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 142.55
Percent Purity ≥90%
Chemical Name or Material Pyrazine-2-carbonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 243-367-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only