missing translation for 'onlineSavingsMsg'
Learn More

Pyrazinamide 98.0+%, TCI America™

Catalog No. P063325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P063325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P063325G Supplier TCI America Supplier No. P063325G

Chemical Identifiers

CAS 98-96-4
Molecular Formula C5H5N3O
Molecular Weight (g/mol) 123.115
MDL Number MFCD00006132
InChI Key IPEHBUMCGVEMRF-UHFFFAOYSA-N
Synonym pyrazinamide, pyrazinecarboxamide, zinamide, pyrazinoic acid amide, 2-pyrazinecarboxamide, aldinamide, aldinamid, pirazinamid, pyrazineamide, pyrafat
PubChem CID 1046
ChEBI CHEBI:45285
IUPAC Name pyrazine-2-carboxamide
SMILES C1=CN=C(C=N1)C(=O)N

Specifications

Melting Point 190°C
Color White
Quantity 25 g
Formula Weight 123.12
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Pyrazinamide
RTECSNumber UQ2275000
TSCA Yes